missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R1 Antibody (OTI2D10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71946AF488
This item is not returnable.
View return policy
Description
CYB5R1 Monoclonal specifically detects CYB5R1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.
Specifications
| CYB5R1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYB5R1 | |
| Full length recombinant protein of human CYB5R1 (NP_057327) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Western Blot | |
| OTI2D10 | |
| Western Blot | |
| b5R.1, B5R1, CYB5BR1, cytochrome b5 reductase 1, EC 1.6.2.2, Humb5R2, NAD(P)H:quinone oxidoreductase type 3 polypeptide A2, NQO3A2polypeptide A2 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 51706.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction