Learn More
CYB5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15469-1-AP-20UL
 View more versions of this product
Product Code. 16814664
  • £135.00 / 20µL
Description
CYB5B gene ontology annotations related to this gene include enzyme activator activity; heme binding; metal ion binding.
Specifications
Specifications
CYB5B
Polyclonal
Unconjugated
CYB5B
CYB5 M, CYB5B, CYB5M, CYPB5M, Cytochrome b5 type B, OMB5
Rabbit
Antigen Affinity Chromatography
RUO
66427, 80773, 80777
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43169, P04166, Q9CQX2
Cyb5b
CYB5B Fusion Protein Ag7771
20 μL
Primary
Rat, Mouse, Human