missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15469-1-AP-150UL
This item is not returnable.
View return policy
Description
CYB5B gene ontology annotations related to this gene include enzyme activator activity; heme binding; metal ion binding.Specifications
| CYB5B | |
| Polyclonal | |
| Unconjugated | |
| CYB5B | |
| CYB5 M, CYB5B, CYB5M, CYPB5M, Cytochrome b5 type B, OMB5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66427, 80773, 80777 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43169, P04166, Q9CQX2 | |
| Cyb5b | |
| CYB5B Fusion Protein Ag7771 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction