missing translation for 'onlineSavingsMsg'
Learn More

CYB561D2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18349755 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349755 25 μg 25µL
18361314 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18349755 Supplier Novus Biologicals Supplier No. NBP31723725UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CYB561D2 Polyclonal antibody specifically detects CYB561D2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB561D2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias cytochrome b-561 domain containing 2,101F6cytochrome b561 domain-containing protein 2, putative tumor suppressor 101F6, Putative tumor suppressor protein 101F6, TSP10
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ILHKEQLGKAHLVTRHGQAGL
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11068
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.