missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYB561D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567642
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
CYB561D1 gene ontology (GO) annotations include oxidation-reduction process.
Specifications
| CYB561D1 | |
| Polyclonal | |
| Unconjugated | |
| CYB561D1 | |
| 1600010M23Rik; AI605489; AW457162; Cyb561d1; cytochrome b-561 domain containing 1; cytochrome b561 domain-containing protein 1; cytochrome b561 family member D1; cytochrome b561 family, member D1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284613, 362017, 72023 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2AE42, Q8N8Q1 | |
| CYB561D1 | |
| Synthesized peptide derived from human CYB561D1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction