missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric Chloride, TRC

Product Code. 30136935
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136935 500 g 500g
30009449 5 g 5g
30003889 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136935 Supplier Toronto Research Chemicals Supplier No. TRCC987695500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.41
Synonym 1,3,5-Trichloro-2,4,6-triazine, 1,3,5-Trichlorotriazine, 2,4,6-Trichloro-1,3,5-triazine, 2,4,6-Trichloro-s-triazine, 2,4,6-Trichloro-sym-triazine, 2,4,6-Trichlorotriazine, Cyanur Chloride, Cyanuric Chloride, Cyanuric Trichloride, Isocyanuric Trichloride, NSC 3512, Solgel W 08, Trichloro-s-triazine, Trichlorocyanidine, Zorugeru W 08, s-Triazine Trichloride, sym-Trichlorotriazine;
IUPAC Name 2,4,6-trichloro-1,3,5-triazine
SMILES Clc1nc(Cl)nc(Cl)n1

Tekniske data

Quantity 500 g
InChI Formula InChI=1S/C3Cl3N3/c4-1-7-2(5)9-3(6)8-1
Formula Weight 182.9158
Chemical Name or Material Cyanuric Chloride
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.