missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric acid, 99%

Product Code. 11492797
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492797 250 g 250g
11374539 100 g 100g
11482797 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492797 Supplier Thermo Scientific Alfa Aesar Supplier No. A15447.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-80-5 | C3H3N3O3 | 129.08 g/mol

Cyanuric acid acts as a chlorine stabilizer in swimming pools. It is used as a precursor to N-chlorinated cyanurates, which are used to disinfect water and crosslinking agents for polyurethane resins. It serves as a process regulator. Further, it is used in the diagnostic determination of melamine and related compounds in kidney tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-80-5
Molecular Formula C3H3N3O3
Molecular Weight (g/mol) 129.08
MDL Number MFCD00082990
InChI Key ZFSLODLOARCGLH-UHFFFAOYSA-N
Synonym cyanuric acid, isocyanuric acid, 1,3,5-triazine-2,4,6-triol, trihydroxycyanidine, tricyanic acid, sym-triazinetriol, s-triazinetriol, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, pseudocyanuric acid, isocyanurate acid
PubChem CID 7956
ChEBI CHEBI:38028
IUPAC Name 1,3,5-triazinane-2,4,6-trione
SMILES O=C1NC(=O)NC(=O)N1

Specifications

Melting Point >360°C
Assay Percent Range 99%
Beilstein 126982
Merck Index 14,2698
Solubility Information Soluble in sulfuric acid,dimethylformamide,sodium hydroxide,potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone,benzene,diethyl ether,ethanol and hexane. Insoluble in cold methanol,benzene and chloroform.
Formula Weight 129.08
Percent Purity 99%
Density 2.5
Quantity 250 g
Chemical Name or Material Cyanuric Acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.