missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric Acid (1.0 mg/10 mL in 84:16% ACN:H2O), TRC

Product Code. 30400660
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400660 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400660 Supplier Toronto Research Chemicals Supplier No. TRCKIT15055X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

CAS 108-80-5
Molecular Formula C3 H3 N3 O3
Quantity 5 x 1 mL
Synonym Isocyanuric acid (6CI), s-Triazine-2,4,6(1H,3H,5H)-trione (8CI), 1,3,5-Triazine-2,4,6-trione, 2,4,6-Trihydroxy-1,3,5-triazine, 2,4,6-Trihydroxy-s-triazine, 2,4,6-Trioxohexahydro-1,3,5-triazine, 5-Azabarbituric acid, Cyanuric acid, ICA-P, NSC 6284, Pseudocyanuric acid, Tricyanic acid, Trihydroxycyanidine, s-Triazine-2,4,6-triol
InChI Formula InChI=1S/C3H3N3O3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)
SMILES O=C1NC(=O)NC(=O)N1
IUPAC Name 1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol) 129.07
Formula Weight 129.0174
Concentration or Composition (by Analyte or Components) Cyanuric acid 1 mg/mL
Chemical Name or Material Cyanuric acid
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.