missing translation for 'onlineSavingsMsg'
Learn More

CyanoguanidineMetformin Related Compound A USP, TRC

Product Code. 17979084
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17979084 5 g 5g
17919454 1 g 1g
17927354 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17979084 Supplier Toronto Research Chemicals Supplier No. TRCC9813505G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2 H4 N4
Molecular Weight (g/mol) 84.08
Synonym Cyanoguanidine, Metformin Hydrochloride Imp. A (EP), Dicyanodiamide, Guanidine, cyano- (8CI,9CI), N-Cyanoguanidine, 1-Cyanoguanidine, 100SF, 1400F, 2-Cyanoguanidine, 200X, 3636AS, AB 04, ACR-H 3636, ACR-H 3636AS, AH 150, AH 154, AH 162, Adeka EH 3636, Adeka EH 3636AS, Adeka ES 3636, Adeka HT 2844, Adeka Hardener 3636AS, Adeka Hardener EH 3636, Adeka Hardener EH 3636AS, Adeka Hardener ESES 3636AS, Ajicure AH 150, Amicure 1400, Amicure 3809, Amicure AH 150, Amicure AH 154, Amicure AH 162, Amicure CG 1200, Amicure CG 1200E, Amicure CG 1200G, Amicure CG 140, Amicure CG 1400, Amicure CG 200, Amicure CG 325, Araldite HT 986, Araldite XB 2879B, Araldite XB 2979B, Bakelite EPH 714, Bakelite VE 2560, Butan 7816LT, CG, CG 1200, CG 1200G, CG 1400, CG 1400S, CGNA, CL 310, Casamid 780, Cyanoguanidine, DCD, DCDA, DDA, DDA (hardener), DDA 10, DDA 5, DDA 50, DICY 1400, DICY 15, DICY 7, DICY 70, DICY 7A, DICY 7S, DICY 7T, DICY-FP, DSH 100, Dicy, Dicy 100S, Dicyandiamide, Dicyanex 1400B, Dicyanex 1400F, Dicyanex 1400HF, Dicyanex 200, Dicyanex 200X, Dicyanex 325, Dicyanodiamide, Didin, Dyhard 100, Dyhard 100E, Dyhard 100M, Dyhard 100S, Dyhard 100SF, Dyhard 100SH, Dyhard D 50EP, Dyhard DF 50EP, Dyhard III, Dyhard PI Optional, Dyhard RU 100, Dyhard SF, Dyhard SF 100, EH 3636AS, EH 3636S, EH 4351S, EPH 714, EQ 14, Ecure 14, Epicure DICY, Epicure DICY 15, Epicure DICY 50, Epicure DICY 7, Epicure DICY 7A, Epicure DICY 7MD, Epicure DICY 7S, Epikure DICY 15, Epikure DICY 7, Erisys DDA 10, GC 120, H 106, H 106 (hardener), H 3636S, HTP 302, Hardener 3636AS, JER Cure DDA 7, JER Cure DICY 15, JER Cure DICY 20, JER Cure DICY 3, JER Cure DICY 7, JER Cure DICY 7T, NSC 2031, Omicure DDA 10, Omicure DDA 5, Omicure DDA 50, SA 284, SF 100, SKW 100S, SKW 100SF, SPX 7922, Technicure D 10, Technicure NanoDicy, Unifill B, XB 2879B, eco-n, jER DICY 20
IUPAC Name 1-cyanoguanidine
SMILES NC(=N)NC#N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)
Formula Weight 84.0436
Chemical Name or Material Cyanoguanidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.