missing translation for 'onlineSavingsMsg'
Learn More

Cyanogen bromide

Product Code. 15411287
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411287 5 g 5g
15421287 25 g 25g
15431287 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411287 Supplier Thermo Scientific Alfa Aesar Supplier No. 032731.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-68-3 | CBrN | 105.922 g/mol

Cyanogen bromide is used as a reagent in molecular biology, peptide mapping, gold extraction and amino acid sequence analysis. It acts as a coupling reagent for phosphoramidate or pyrophosphate inter-nucleotide bonds in DNA duplexes. It is also used in organic synthesis to prepare cyanamides, dicyanamides, cyanuric bromide, guanidines and hydroxylguanidines. It is involved in the von Braun reaction, it gives disubstituted cyanamides by reacting with tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-68-3
Molecular Formula CBrN
Molecular Weight (g/mol) 105.922
MDL Number MFCD00011597
InChI Key ATDGTVJJHBUTRL-UHFFFAOYSA-N
Synonym cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
PubChem CID 10476
IUPAC Name carbononitridic bromide
SMILES C(#N)Br

Specifications

Melting Point 50°C to 52°C
Density 2.015
Boiling Point 61.4°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethanol,diethyl ether,benzene and acetonitrile.
Formula Weight 105.93
Physical Form Crystals
Chemical Name or Material Cyanogen bromide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyanogen bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Cyanogen bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.