missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetic acid, 97%

Product Code. 10273940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
250g
25g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10273940 1 kg Glass bottle 1kg
10426910 25 g Glass bottle 25g
10436910 250 g Glass bottle 250g
11985601 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10273940 Supplier Thermo Scientific Acros Supplier No. 110670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-09-8 | C3H3NO2 | 85.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-09-8
Molecular Formula C3H3NO2
Molecular Weight (g/mol) 85.06
InChI Key MLIREBYILWEBDM-UHFFFAOYSA-N
Synonym cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova
PubChem CID 9740
ChEBI CHEBI:51889
IUPAC Name 2-cyanoacetic acid
SMILES C(C#N)C(=O)O

Specifications

Melting Point 65.0°C to 67.0°C
Color White to Beige
Boiling Point 108.0°C (15.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NCCH2CO2H
Quantity 1 kg
Beilstein 02,583
Fieser 11,145; 13,171
Merck Index 15,2679
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene and chloroform
Formula Weight 85.06
Percent Purity 97%
Physical Form Adhering Crystalline Solid
Chemical Name or Material Cyanoacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.