missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyano-o-tolunitrile, 80%, Thermo Scientific™

Product Code. 10418690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10418690 100 g Glass bottle 100g
10021680 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10418690 Supplier Thermo Scientific Acros Supplier No. 152571000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3759-28-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001895
InChI Key GKHSEDFDYXZGCG-UHFFFAOYSA-N
Synonym 2-cyanomethyl benzonitrile, homophthalonitrile, 2-cyanobenzyl cyanide, 2-cyanophenylacetonitrile, alpha-cyano-o-tolunitrile, o-cyanobenzyl cyanide, benzeneacetonitrile, 2-cyano, 2-cyanobenzeneacetonitrile, o-cyanophenyl acetonitrile, o-tolunitrile, alpha-cyano
PubChem CID 77368
IUPAC Name 2-(cyanomethyl)benzonitrile
SMILES N#CCC1=CC=CC=C1C#N

Specifications

CAS Min % 75.0
CAS Max % 100.0
Melting Point 76°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Linear Formula NCCH2C6H4CN
Quantity 100 g
Beilstein 09, 859
Formula Weight 142.16
Percent Purity 80%
Grade Technical
Physical Form Chunks and/or Powder
Chemical Name or Material α-Cyano-o-tolunitrile, tech.
Show More Show Less
compliance-icons
Product Identifier
  • a-Cyano-o-tolunitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.