missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cyanine 5, SE

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Cyanine 5, SE is a red fluorescent dye for the labeling of amines. Excitation maximum ∽649 nm; emission maximum ∽666 nm.
Specifications
Specifications
| Chemical Name or Material | 2-[5-[1-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-1,3-dihydro-3,3-dimethyl-5-sulfo-2 H-indol-2-ylidene]-1,3-pentadien-1-yl]-1-ethyl-3,3-dimethyl-5-sulfo-3 H-indolium, inner salt |
| CAS | 146368-14-1 |
| Synonym | Cy5 NHS Ester |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in DMF |
| Target | Fluorescent Dyes |
| Molecular Formula | C37H43N3O10S2 |
| Formula Weight | 753.88 |
| Physical Form | Solid |
| Color | White |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction