missing translation for 'onlineSavingsMsg'
Learn More

CY 208-243, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001216
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001216

Brand: Tocris Bioscience 1249/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective D1-like agonist

Centrally active dopamine D1 receptor agonist, selective over D2 receptor sites. Stimulates adenylate cyclase in rat striatal homogenates with an EC50 of 125 nM. Unlike SKF 38393 (Cat. No. 0922), it exerts antiParkinsonian activity in animal models.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100999-26-6
Molecular Formula C19H18N2
Molecular Weight (g/mol) 274.367
InChI Key WRNKIDLXXXIELU-IEBWSBKVSA-N
Synonym indolophenanthridine, 4,6,6a,7,8,12b-hexahydro-7-methylindolo 4,3-ab-phenanthridine, indolo 4,3-ab phenanthridine, 4,6,6a,7,8,12b-hexahydro-7-methyl-, trans--, 6ar,12br-7-methyl-4,6,6a,7,8,12b-hexahydroindolo 4,3-ab phenanthridine, tocris-1249, biomol-nt_000032, d0bq8m, d0f5vr, --6ar,12br-4,6,6a,7,8,12b-hexahydro-7-methylindolo 4,3-a phenanthridin
PubChem CID 58144
SMILES CN1CC2=CC=CC=C2C3C1CC4=CNC5=CC=CC3=C45

Specifications

Quantity 10 mg
Formula Weight 274.36
Percent Purity >98%
Chemical Name or Material CY 208-243
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.