missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CY 09
Description
CY 09 is a NLRP3 inhibitor (Kd = 500 nM); directly binds to ATP-binding motif of NLRP3 NACHT domain. Blocks NLRP3 inflammasome assembly and activation. Displays therapeutic effects on mouse models of cryopyrin-associated autoinflammatory syndrome (CAPS) and type 2 diabetes. Exhibits no activity for hERG (10 μM).
Specifications
Specifications
| Chemical Name or Material | 4-[[4-Oxo-2-thioxo-3-[[3-(trifluoromethyl)phenyl]methyl]-5-thiazolidinylidene]methyl]benzoic acid |
| CAS | 1073612-91-5 |
| Quantity | 10 mg |
| Molecular Formula | C19H12F3NO3S2 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?