missing translation for 'onlineSavingsMsg'
Learn More

CY-09, MedChemExpress

Product Code. 16476607
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476607 5 mg 5mg
16471029 1 mg 1mg
16470247 50 mg 50mg
16444926 100 mg 100mg
16423888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476607 Supplier Medchem Express Supplier No. HY103666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CY-09 is a selective and direct NLRP3 inhibitor. CY-09 directly binds to the ATP-binding motif of NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, resulting in the suppression of NLRP3 inflammasome assembly and activation.

CY-09 is a selective and direct NLRP3 inhibitor. CY-09 directly binds to the ATP-binding motif of NLRP3 NACHT domain and inhibits NLRP3 ATPase activity, resulting in the suppression of NLRP3 inflammasome assembly and activation.

Specifications

Chemical Name or Material CY-09
Physical Form Solid
CAS 1073612-91-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H12F3NO3S2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 150 mg/mL (354.25 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC(C1=CC=C(/C=C2SC(N(CC3=CC=CC(C(F)(F)F)=C3)C\2=O)=S)C=C1)=O
Molecular Weight (g/mol) 423.43
Formula Weight 423.43
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.