missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXXC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16513-1-AP-20UL
This item is not returnable.
View return policy
Description
CXXC5 is zinc finger protein closely related to the DVL1-binding protein CXXC4 (also known as Idax), and like CXXC4 is thought to modulate the Wnt signaling pathway during development. CXXC5 is upregulated in neural stem cells upon BMP4 stimulation, leading to decreased levels of the Wnt-signaling target AXIN2 and facilitated the response of the stem cells to Wnt3a. Recent reports suggest CXXC5 transcription is activated by the Wilms tumor 1 (WT1) transcription factor, and together are involved in the WNT/beta-catenin pathway and play a role in embryonic kidney development.Specifications
| CXXC5 | |
| Polyclonal | |
| Unconjugated | |
| CXXC5 | |
| CXXC finger 5, CXXC5, HSPC195, TCCCIA00297 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291670, 51523, 67393 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XIQ3, Q7LFL8, Q91WA4 | |
| Cxxc5 | |
| CXXC5 Fusion Protein Ag9733 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction