missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXXC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27963-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.Specifications
| CXXC1 | |
| Polyclonal | |
| Unconjugated | |
| CXXC1 | |
| 2410002I16Rik, 5830420C16Rik, CFP1, CGBP, CpG binding protein, CXXC finger 1 (PHD domain), CXXC1, hCGBP, HsT2645, PCCX1, PHF18, SPP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30827 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9P0U4 | |
| CXXC1 | |
| CXXC1 Fusion Protein Ag27558 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction