missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXXC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92690-0.02ml
This item is not returnable.
View return policy
Description
CXXC1 Polyclonal antibody specifically detects CXXC1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CXXC1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:20 - 1:50 | |
| CFP1CXXC-type zinc finger protein 1, CGBPPHD finger and CXXC domain-containing protein 1, CpG binding protein, cpG-binding protein, CXXC finger 1 (PHD domain), CXXC finger protein 1, DNA-binding protein with PHD finger and CXXC domain, hCGBP, HsT2645, PCCX12410002I16Rik, PHF185830420C16Rik, SPP1, ZCGPC1, zinc finger, CpG binding-type containing 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human CXXC1 (NP_001095124.1). MEGDGSDPEPPDAGEDSKSENGENAPIYCICRKPDINCFMIGCDNCNEWFHGDCIRITEKMAKAIREWYCRECREKDPKLEIRYRHKKSRERDGNERDSSEPRDEGGGRKRPVPDPDLQRRAGSGTGVGAMLARGSASPHKSSPQPLVATPSQHHQQQQQQIKRSARMCGECEACRRTEDCGHCDFCRDMKKFGGPNKIRQKCRLRQCQL | |
| 0.02 mL | |
| Neuroscience, Unfolded Protein Response (UPR) | |
| 30827 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction