missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CXorf66 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569334
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
The protein encoded by this gene is predicted to be a type I membrane protein, however, its exact function is not known.
Specifications
| CXorf66 | |
| Polyclonal | |
| Unconjugated | |
| CXorf66 | |
| chromosome X open reading frame 66; CXorf66; RP11-35F15.2; secreted glycoprotein, X-linked; SGPX; Uncharacterized protein CXorf66 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 347487 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5JRM2 | |
| CXorf66 | |
| synthetic peptide directed towards the middle region of human CXorf66. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion