missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CXorf36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546135
This item is not returnable.
View return policy
Description
Peptide sequence: DKQEGSQEAN RAGENKDIFS CLVSGCQAQL PSCESISEKQ SLVLVCQKLL Sequence homology: Horse: 79%; Human: 100%; Mouse: 79%.
The function of this protein remains unknown.
Specifications
| CXorf36 | |
| Polyclonal | |
| Unconjugated | |
| CXorf36 | |
| 4930578C19Rik; bA435K1.1; chromosome X open reading frame 36; CXorf36; deleted in autism-1 related protein; deleted in autism-related protein 1; DIA1R; DIPK2B; Divergent protein kinase domain 2B; EPQL1862; hCG1981635; PRO3743; UNQ1862/PRO3743; UPF0672 protein CXorf36 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79742 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H7Y0 | |
| CXorf36 | |
| Synthetic peptide directed towards the C-terminal of human CXorf36 (aa 317-366). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction