missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CXorf22 Recombinant Protein Antigen

Product Code. 18474892 Shop All Bio Techne Products
Click to view available options
:
0.1 mL
This item is not returnable. View return policy

Product Code. 18474892

missing translation for 'mfr': Novus Biologicals™ NBP230729PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 286464
Molecular Weight (g/mol) 28 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias Calponin Homology Domain Containing 2, Calponin Homology Domain-Containing Protein 2, CHDC2, chromosome X open reading frame 22, Chromosome X Open Reading Frame 30, chromosome X open reading frame 59, Cilia And Flagella Associated Protein 47, Cilia- And Flagella-Associated Protein 47, CXorf30, CXorf59, MGC34831
Gene Symbol CFAP47
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-30729. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CXorf22
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.