missing translation for 'onlineSavingsMsg'
Learn More

CXD101, MedChemExpress

Product Code. 16475887
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475887 5 mg 5mg
16459706 50 mg 50mg
16433158 10 mg 10mg
16410699 1 mg 1mg
16444446 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16475887 Supplier Medchem Express Supplier No. HY100748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CXD101 is a potent, selective and orally active class I HDAC inhibitor with IC50s of 63 nM, 570 nM and 550 nM for HDAC1, HDAC2 and HDAC3, respectively. CXD101 has no activity against HDAC class II. CXD101 has antitumor activity.

CXD101 is a potent, selective and orally active class I HDAC inhibitor with IC50s of 63 nM, 570 nM and 550 nM for HDAC1, HDAC2 and HDAC3, respectively. CXD101 has no activity against HDAC class II. CXD101 has antitumor activity.

Specifications

Chemical Name or Material CXD101
CAS 934828-12-3
Color White
Purity Grade Notes Research
Molecular Formula C24H29N5O
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (76.82 mM)
SMILES O=C(NC1=C(N)C=CC=C1)C2=CC=C(C3CCN(CC4=CN(C)N=C4C)CC3)C=C2
Molecular Weight (g/mol) 403.52
Formula Weight 403.52
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.