missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7/RDC-1 Rabbit anti-Human, Mouse, Rat, Clone: S08-6H8, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | CXCR7/RDC-1 |
|---|---|
| Clone | S08-6H8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18378824
|
Novus Biologicals
NBP3-14920-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18300543
|
Bio-Techne
NBP3-14920-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR7/RDC-1 Monoclonal antibody specifically detects CXCR7/RDC-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CXCR7/RDC-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, GPCR | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 57007 | |
| IgG | |
| Affinity-purified |
| S08-6H8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| A synthetic peptide of human CXCR7/RDC-1 (Uniprot#: P25106) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title