missing translation for 'onlineSavingsMsg'
Learn More

CXCR7/RDC-1 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18431515 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431515 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431515 Supplier Novus Biologicals Supplier No. NBP224779IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CXCR7/RDC-1 Polyclonal antibody specifically detects CXCR7/RDC-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR7/RDC-1
Applications Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding amino acids 106-129 of human CXCR7/RDC1 was used as the immunogen, GenBank NP_064707.1.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57007
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.