missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CXCR7 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This antibody recognizes endogenous CXCR7, which has an experimentally determined molecular weight of 50 kDa.
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.Specifications
| CXCR7 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| O89039, P25106, P56485 | |
| 12778, 57007, 84348 | |
| CXCR7 Fusion Protein Ag14247 | |
| Primary | |
| -20°C | |
| ACKR3 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Chemokine orphan receptor 1, CMKOR1, CXC R7, CXCR 7, CXCR7, G protein coupled receptor 159, GPR159, RDC 1, RDC1 | |
| ACKR3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title