missing translation for 'onlineSavingsMsg'
Learn More

CXCR7 modulator 2, MedChemExpress

Product Code. 16457297
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457297 5 mg 5mg
16420887 50 mg 50mg
16415436 100 mg 100mg
16404588 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16457297 Supplier Medchem Express Supplier No. HY112154
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CXCR7 modulator 2 is a modulator of C-X-C Chemokine Receptor Type 7 (CXCR7), with a Ki of 13 nM.

CXCR7 modulator 2 is a modulator of C-X-C Chemokine Receptor Type 7 (CXCR7), with a Ki of 13 nM.

Specifications

Chemical Name or Material CXCR7 modulator 2
CAS 2227426-37-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C29H42N6O3
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (478.30 mM; Need ultrasonic)
SMILES O=C([C@@H]1[C@H](O2)CC[C@H]2C1)N3CCC([C@H](CC(N)=O)N4CCCN(C5=C(CC)C=CN6C5=NC=C6)CC4)CC3
Molecular Weight (g/mol) 522.68
Formula Weight 522.68
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.