Learn More
CXCR5 Rat anti-Mouse, PE, Clone: 2G8, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 551959
Description
The monoclonal antibody 2G8 reacts with the mouse CXC chemokine receptor, CXCR5. CXCR5 (a.k.a. BLR1, NLR and MDR15), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CXC chemokines, CXCL13/BLC/BCA-1. The expression of CXCR5 has been detected in spleen, lymph nodes, tonsils, brain, bone marrow, T cells, B cells, cerebrum, cerebellum, hippcampus and pituitary. In mouse spleen, CXCR5 was strictly expressed by mature B cells and a small subset of T lymphocytes. The immunogen used to generate 2G8 hybridoma was a recombinant protein containing N-terminal amino acids of mouse CXCR5 (GST-NmBLR1). This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 2G8
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse CXCR5
Flow Cytometry
Specifications
| CXCR5 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CXCR5 | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 2G8 | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.