missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
£186.00 - £419.00
Specifications
| Antigen | CXCR4 |
|---|---|
| Dilution | Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18705073
|
Novus Biologicals
NB100-74396SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706383
|
Novus Biologicals
NB100-74396 |
£419.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CXCR4 Polyclonal specifically detects CXCR4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CXCR4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
| Whole antisera with 0.05% Sodium Azide | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
| CXCR4 | |
| IgG | |
| Unpurified |
| Western Blot 1:5000, Flow Cytometry, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| P70658 | |
| 7852 | |
| A synthetic peptide made to a C-terminal region of the human CXCR4 protein (within residues 300-352). [Swiss-Prot P61073] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title