missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CXCR3 Antibody (Genzyme patent anti-CXCR3) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28371-1mg
This item is not returnable.
View return policy
Description
CXCR3 Monoclonal antibody specifically detects CXCR3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXCR3 | |
| Monoclonal | |
| Unconjugated | |
| P49682 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genzyme patent anti-CXCR3 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| CXCR3/ GPR9/ CD183 | |
| 1 mg | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| 2833 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction