missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR3 Antibody (5C10E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £466.00
Specifications
| Antigen | CXCR3 |
|---|---|
| Clone | 5C10E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18022826
|
Novus Biologicals
NBP2-61681 |
0.1 mL |
£466.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698776
|
Novus Biologicals
NBP2-61681-0.025ml |
0.025 mL |
£184.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR3 Monoclonal specifically detects CXCR3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CXCR3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| CXCR3 | |
| IgG1 | |
| Protein G purified |
| 5C10E6 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| 2833 | |
| Purified recombinant fragment of human CD183 (AA: extra mix) expressed in E. Coli. | |
| Primary | |
| 40.7 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title