missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20634-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. This receptor also binds to chemokine (C-X-C motif) ligand 1 (CXCL1/MGSA), a protein with melanoma growth stimulating activity, and has been shown to be a major component required for serum-dependent melanoma cell growth. This receptor mediates neutrophil migration to sites of inflammation. The angiogenic effects of IL8 in intestinal microvascular endothelial cells are found to be mediated by this receptor. Knockout studies in mice suggested that this receptor controls the positioning of oligodendrocyte precursors in developing spinal cord by arresting their migration. This gene, IL8RA, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36. Alternatively spliced variants, encoding the same protein, have been identified.Specifications
| CXCR2 | |
| Polyclonal | |
| Unconjugated | |
| CXCR2 | |
| CD182, CDw128b, CMKAR2, CXC R2, CXCR 2, CXCR2, GRO/MGSA receptor, IL 8 receptor type 2, IL 8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor, beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3579 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P25025 | |
| CXCR2 | |
| CXCR2 Fusion Protein Ag14670 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction