Learn More
Invitrogen™ CXCR2 Monoclonal Antibody (48311.211)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA124669
Product Code. 11564752
  • £439.00 / 100µg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCR2
Monoclonal
0.5 mg/mL
PBS with no preservative
P25025
CXCR2
Human CXCR2 transfected NSO mouse myeloma cells.
100 μg
Primary
Human
Antibody
IgG2a
Flow Cytometry, Inhibition Assays, Neutralization
48311.211
Unconjugated
CXCR2
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; CD128; CD182; CD182 antigen; CDw128; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (CXC) receptor 2; chemokine (C-X-C) receptor 2; Cmkar2; CXC; C-X-C chemokine receptor type 2; CXC motif chemokine; C-X-C motif chemokine receptor 2; Cxcr2; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; Gpcr16; GRO/MGSA receptor; high affinity interleukin-8 receptor B; IL-8 receptor alpha chain; Il-8 receptor b; IL-8 receptor beta; IL-8 receptor type 2; IL-8R B; IL8R2; IL8RA; Il8rb; IL-8rb; IL-8Rh; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B; mIL-8RH
Mouse
Protein G
RUO
3579
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.