missing translation for 'onlineSavingsMsg'
Learn More

CXCR1/IL-8RA Antibody (SE2), DyLight 488, Novus Biologicals™

Product Code. 18355602 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355602 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355602 Supplier Novus Biologicals Supplier No. NBP312003G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CXCR1/IL-8RA Monoclonal antibody specifically detects CXCR1/IL-8RA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR1/IL-8RA
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SE2
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Host Species Rabbit
Immunogen This antibody was raised by immunizing BALB/c mice with a BSA-conjugated peptide comprising of the N-terminal part of CXCR1/IL-8RA (amino acids 1-30).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3577
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.