missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL9/MIG Antibody (Novimmune patent anti-CXCL9) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28413JF585
This item is not returnable.
View return policy
Description
CXCL9/MIG Monoclonal antibody specifically detects CXCL9/MIG in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXCL9/MIG | |
| Monoclonal | |
| Janelia Fluor 585 | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| CXCL9 | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
| 4283 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Novimmune patent anti-CXCL9 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction