missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL9/MIG Antibody (D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43439
This item is not returnable.
View return policy
Description
CXCL9/MIG Monoclonal antibody specifically detects CXCL9/MIG in Human samples. It is validated for Western Blot
Specifications
| CXCL9/MIG | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| Immunized with recombinant Human CXCL9/MIG (Uniprot#: Q07325; Thr23∽Thr125) | |
| 100 μL | |
| Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
| 4283 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot | |
| D1 | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction