missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | CXCL9/MIG |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 277 |
| Conjugate | Alexa Fluor 594 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CXCL9/MIG (P18340; Thr22-Thr126). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?