missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL9/MIG Antibody (050), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89697AF594
This item is not returnable.
View return policy
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| CXCL9/MIG | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Mouse CXCL9 Cynomolgus CXCL9 No cross-reactivity in ELISA with: Human CXCL10 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 50 | |
| ELISA, Sandwich ELISA Detection | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CXCL9/MIG (Q07325; Thr23-Thr125). | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
| 4283 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction