missing translation for 'onlineSavingsMsg'
Learn More

CXCL8/IL-8 Mouse anti-Bovine, Canine, Clone: MT8H6, Novus Biologicals™

Product Code. 18337926 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1000 μg
250 μg
Unit Size:
1000µg
250µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337926 250 μg 250µg
18310894 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18337926 Supplier Novus Biologicals Supplier No. NBP318279250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Bovine, Canine samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL8/IL-8
Applications ELISA
Classification Monoclonal
Clone MT8H6
Concentration 0.5 mg/ml
Conjugate Unconjugated
Formulation 0.2 um filtered PBS
Gene Alias 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor
Host Species Mouse
Immunogen Recombinant human CXCL8/ IL-8
Purification Method Protein G purified
Quantity 250 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3576
Target Species Bovine, Canine
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.