missing translation for 'onlineSavingsMsg'
Learn More

CXCL7/NAP-2 Antibody (3B9), Novus Biologicals™

Product Code. 18324769 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324769 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324769 Supplier Novus Biologicals Supplier No. H00005473M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL7/NAP-2 Monoclonal antibody specifically detects CXCL7/NAP-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL7/NAP-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Sandwich ELISA, KnockDown
Classification Monoclonal
Clone 3B9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Sandwich ELISA, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH28217
Gene Alias Beta-TG, connective tissue-activating peptide III, CTAP3CTAP-III, CTAPIII, CXC chemokine ligand 7, C-X-C motif chemokine 7, CXCL7b-TG1, LA-PF4, LDGFTC1, Leukocyte-derived growth factor, low-affinity platelet factor IV, Macrophage-derived growth factor, MDGFTC2, NAP-2, NAP-2-L1, neutrophil-activating peptide 2, neutrophil-activating peptide-2, PBPTHBGB, platelet basic protein, pro-platelet basic protein (chemokine (C-X-C motif) ligand 7), SCAR10, SCYB7beta-thromboglobulin, small inducible cytokine B7, small inducible cytokine subfamily B, member 7, Small-inducible cytokine B7, TGB, TGB1B-TG1, THBGB1, thrombocidin 1, thrombocidin 2, thromboglobulin, beta-1
Host Species Mouse
Immunogen PPBP (AAH28217, 37 a.a. ~ 128 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TKGQTKRNLAKGKEESLDSDLYAELRCMCIKTTSGIHPKNIQSLEVIGKGTHCNQVEVIATLKDGRKICLDPDAPRIKKIVQKKLAGDESAD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5473
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.