missing translation for 'onlineSavingsMsg'
Learn More

CXCL6/GCP-2 Antibody (MM0217-10L25), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18429115 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429115 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429115 Supplier Novus Biologicals Supplier No. NBP212232JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL6/GCP-2 Monoclonal antibody specifically detects CXCL6/GCP-2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL6/GCP-2
Applications Western Blot, ELISA
Classification Monoclonal
Clone MM0217-10L25
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2), Chemokine alpha 3, CKA-3Granulocyte chemotactic protein 2, GCP2member 6 (granulocytechemotactic protein 2), GCP-2Small-inducible cytokine B6, Small inducible cytokine subfamily B (Cys-X-Cys), member b
Host Species Mouse
Immunogen Human recombinant CXCL6/GCP-2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6372
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.