Learn More
PeproTech CXCL5 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P91BT-500UG
Description
AA Sequence of recombinant protein: AAVLRELRCV CLQTTQGVHP KMISNLQVFA IGPQCSKVEV VASLKNGKEI CLDPEAPFLK KVIQKILDGG NKEN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human ENA-78 (CXCL5). Anti-Human ENA-78 (CXCL5)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hENA-78 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human ENA-78 (500-P91) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hENA-78. Western Blot: To detect hENA-78 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hENA-78 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P91BT-1MG will be provided as 2 x 500 μg
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
Specifications
| CXCL5 | |
| Polyclonal | |
| Biotin | |
| CXCL5 | |
| AMCF-II; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; CXCL; CXCL5; Cxcl6; Cytokine LIX; ENA78; ENA-78; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; GCP-2; GCP-2(1-78); GCP-2(9-78); granulocyte chemotactic protein-2; LIX; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; Scyb5; Scyb6; small inducible cytokine B subfamily, member 5; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); small inducible cytokine subfamily B, member 15; small-inducible cytokine B5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6374 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P42830 | |
| CXCL5 | |
| E.coli-derived Recombinant Human ENA-78 (CXCL5) (5-78 a.a.). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.