missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL5/ENA-78 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £452.00
Specifications
| Antigen | CXCL5/ENA-78 |
|---|---|
| Clone | 33103 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
18784239
![]() |
R&D Systems
MAB2541-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18151941
![]() |
R&D Systems
MAB2541 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot.Specifikationer
| CXCL5/ENA-78 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
| CXCL5 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 33103 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P42830 | |
| 6374 | |
| E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 Accession # P42830 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CXCL5/ENA-78 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CXCL1, 2, 3, 4, 6, 7, 8, 9, 10, 11, 12, 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12, 15, 16, recombinant rat CXCL5, and recombinant porcine CXCL8 is observed. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel