missing translation for 'onlineSavingsMsg'
Learn More

CXCL5/ENA-78 Antibody (33160) [PerCP], Novus Biologicals™

Product Code. 30732563 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732563 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732563 Supplier Novus Biologicals Supplier No. FAB254C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL5/ENA-78
Applications Immunohistochemistry, Western Blot, Neutralization
Classification Monoclonal
Clone 33160
Conjugate PerCP
Formulation PBS
Gene Alias chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL5/ENA-78, Ala37-Asn114 (predicted), Accession # P42830
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6374
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.