missing translation for 'onlineSavingsMsg'
Learn More

CXCL5/ENA-78 Antibody (32B3), PE, Novus Biologicals™

Product Code. 18416301 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416301 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416301 Supplier Novus Biologicals Supplier No. NBP222026PE

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL5/ENA-78
Applications Western Blot, ELISA, Neutralization
Classification Monoclonal
Clone 32B3
Conjugate PE
Dilution Western Blot, ELISA, Block/Neutralize
Formulation PBS
Gene Alias chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
Host Species Rat
Immunogen Mouse recombinant CXCL5/ENA-78
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6374
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.