missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Neutralization
Specifications
Specifications
| Antigen | CXCL5/ENA-78 |
| Applications | Western Blot, ELISA, Neutralization |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA, Block/Neutralize |
| Formulation | 50mM Sodium Borate |
| Gene Alias | chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?