Learn More
PeproTech CXCL3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
£177.00 - £2745.00
Specifications
| Antigen | CXCL3 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17854403
![]() |
PeproTech
500-P105BT-25UG |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17853833
![]() |
PeproTech
500-P105BT-50UG |
50 μg |
£228.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17843813
|
Invitrogen
500-P105BT-1MG |
2 x 500 μg | |||||||
|
30054659
|
PeproTech
500-P105BT-500UG |
500 μg |
£1466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GRO-gamma (CXCL3). Anti-Human GRO-gamma (CXCL3)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human GRO-gamma by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GRO-gamma (500-P105) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GRO-gamma. Western Blot: To detect Human GRO-gamma by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human GRO-gamma is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P105BT-1MG will be provided as 2 x 500 μg
CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils.Specifications
| CXCL3 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| P19876 | |
| 2921 | |
| E.coli-derived Recombinant Human GRO-gamma (CXCL3). | |
| Antigen affinity chromatography | |
| Cxcl3 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2b; MIP2-beta; SCYB3 | |
| Cxcl3 | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.