missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CXCL3 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148971

Product Code. 30533857

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion. All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCL3
Polyclonal
Unconjugated
Cxcl3
C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 3; Cinc2; CINC-2; CINC-2b; CXC; CXC motif chemokine; C-X-C motif chemokine 3; C-X-C motif chemokine ligand 3; CXCL; Cxcl3; cytokine-induced neutrophil chemoattractant 2; cytokine-induced neutrophil chemoattractant-2; Dcip1; Dendritic cell inflammatory protein 1; Gm1960; gro gamma; GRO3; GRO3 oncogene; GROG; GRO-gamma; GRO-gamma(1-73); GRO-gamma(5-73); Growth-regulated protein gamma; macrophage inflammatory protein 2-alpha/beta; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma; MIP2-alpha/beta; MIP2B; MIP-2b; MIP2-beta; SCYB3
Goat
Antigen affinity chromatography
RUO
171551
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Neutralization, Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q10746
Cxcl3
E. coli-derived recombinant rat CXCL3/CINC-2 alpha/beta, Arg33-Leu100.
100 μg
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.