Learn More
PeproTech CXCL2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P104BT-500UG
Description
AA Sequence of recombinant protein: APLATELRCQ CLQTLQGIHL KNIQSVKVKS PGPHCAQTEV IATLKNGQKA CLNPASPMVK KIIEKMLKNG KSN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GRO-beta (CXCL2). Anti-Human GRO-beta (CXCL2)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human GRO-beta (CXCL2) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GRO-beta (CXCL2) (500-P104) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GRO-beta (CXCL2). Western Blot: To detect Human GRO-beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human GRO-beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P104BT-1MG will be provided as 2 x 500 μg
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
Specifications
| CXCL2 | |
| Polyclonal | |
| Biotin | |
| Cxcl2 | |
| C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2a; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO β; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROβ; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 a; MIP 2 alpha; MIP 2 α; Mip2; MIP-2; MIP2 a; MIP2 α; MIP2A; MIP-2a; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2920 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P19875 | |
| Cxcl2 | |
| E.coli-derived Recombinant Human GRO-beta (CXCL2). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.