missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL2/GRO beta/MIP-2/CINC-3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03233-100UL
This item is not returnable.
View return policy
Description
CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Human, Rat samples. It is validated for Western Blot.
Specifications
| CXCL2/GRO beta/MIP-2/CINC-3 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| chemokine (C-X-C motif) ligand 2, CINC-2a, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human CXCL2/GRO beta/MIP-2/CINC-3 (NP_002080.1). | |
| 100 μL | |
| Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
| 2920 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction